Insect Wings, Composite Slide
Insect Wings, Composite, Whole Mount, Unstained
Catalog # 470176-946
Supplier: Avantor
Jump to:
Product Details & Documents
Compare and Contrast Wing Types.
- Identifying features clearly distinguishable
- Rigorous quality control standards
Illustrates four types: membranous (housefly), net-veined (dragonfly), scaled (butterfly), and leathery (grasshopper). Unstained.
Specifications
- Description:Insect Wings, Composite
- Stain:Unstained
- Orientation:Whole Mount