ContactUsLinkComponent
Hydrion A and B Dispenser
Hydrion A and B Dispenser
Catalog # 470203-342
Supplier:  Micro Essential
Hydrion A and B Dispenser
Catalog # 470203-342
Supplier:  Micro Essential
Supplier Number:  150
Specifications
  • Description:
    pH Paper, Hydrion A & B Dispenser, Double Roll
  • Cat. No.:
    470203-342
  • Supplier no.:
    150
Specifications
About this item
Use this paper to cover both the A range (2-4-6-8-10) and the B range (1-3-5-7-9-11) of the pH scale.
Includes: color chart.