ContactUsLinkComponent
Human Rib with Embedded Bullet
Human Rib with Embedded Bullet
Catalog # 470030-038
Small Business Enterprise
undefined
Human Rib with Embedded Bullet
Catalog # 470030-038
Supplier Number:  FO-101
Small Business Enterprise
Specifications
  • Description:
    Human Rib with Embedded Bullet
  • Length:
    8"
  • Cat. No.:
    470030-038
  • Supplier no.:
    FO-101
Specifications
About this item
Captures All of the Osteological and Forensic Details of the Original Specimen
Replica. Help students determine post-mortem injuries and bullet trajectories. Shot from above by a .32 caliber firearm, this right rib of a Caucasian male still has the bullet embedded in it. You can determine from the evidence of new bone growth around the site of the injury that the wound was non-fatal. Also visible is an
example of a post-mortem break to the proximal portion of the rib. Size: 8"L.