Human Karyotypes Slide Set
Catalog # 470183-614
Supplier: Avantor
Specifications
- Type:Cellular Development
- Description:Human Karyotypes Slide Set
- No. of slides:3
- Cat. No.:470183-614
- Supplier no.:WARD470183-614
Specifications
About this item
Illustrates Normal Human Chromosomes and Genetic Dysfunctions
- Identifying features clearly distinguishable
- Rigorous quality control standards
Ordering information: Set includes Human Chromosome smears: Male 46 XY; female 46 XX; Trisomy 21: 47 XY +21. The set also includes a teacher and student activity booklet.