Honeybee Worker Leg, Composite Slide
470181-646
: Avantor
- Description:Honeybee Worker Leg, Composite
- Cat. No.:470181-646
- Supplier no.:WARD470181-646
- Orientation:Whole mount
Illustrates Important Worker Bee Body Parts
- Identifying features clearly distinguishable
- Rigorous quality control standards
One view each of the first, second, and third legs. Shows antenna cleaner, spur, and pollen basket. Unstained.