ContactUsLinkComponent
GEO-logic Hardness Set
GEO-logic Hardness Set
Catalog # 470025-336
Supplier:  Avantor
undefined
GEO-logic Hardness Set
Catalog # 470025-336
Supplier:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Hardness Topic Set (9 Specimens)
  • Cat. No.:
    470025-336
  • Supplier no.:
    WARD470025-336
Specifications
About this item
A Complete Kit to Communicate the Concept of Hardness in Minerals
  • Developed jointly by experienced educators and Ward's® staff geologists
  • Classroom tested for dependability
  • Designed to save valuable planning and preparation time
Students will understand the differences in mineral composition when they use our GEO-logic set to investigate variations in mineral hardness. A teacher's guide and student copymasters are included for the following activities: relative scale of hardness, significance of hardness, and procedure for establishing hardness. The set is housed in a compartmented collection box.
GEO-logic is a unique, comprehensive series of teaching collections designed to assist both teacher and student in understanding basic earth science concepts through hands-on activities and carefully prepared teacher and student guides.
Contents:
9 individually numbered samples, 1×2"
Streak plate
Magnifier
Copper strip
Glass plate
Nail
Minerals: apatite, calcite, corundum, fluorite, microcline, quartz, selenite, talc, topaz
Caution: Due to the nature of this product it may contain asbestos.