Ward's® Live Gregarines Culture
Catalog # 470180-138
Supplier: Avantor
Specifications
- Description:Gregarines (Small)
- No. of students:30
- Cat. No.:470180-138
- Packaging:Jar
- Media:Live mealworms
- Supplier no.:WARD470180-138
Specifications
About this item
Lives in intestine of host — mealworms — as symbiotic organism. Culture of mealworms provided.
Produce small, infective spores.
Ordering information: These products contain living or perishable materials
Delivery: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.