Graph Papers
: IMAGEN
The logarithmic paper has a one cycle semi-log chart.
The metric graph paper is ruled 10 lines per cm with the fifth and tenth line accented. The polar coordinate paper has every degree indicated and every 10° accented. Each is three-hole punched and measures 22×28 cm. Packages of 100.
The logarithmic paper has a one cycle semi-log chart.
The metric graph paper is ruled 10 lines per cm with the fifth and tenth line accented. The polar coordinate paper has every degree indicated and every 10° accented. Each is three-hole punched and measures 22×28 cm. Packages of 100.
Product Family Options
- Description
- Graph paper, metric coordinates, 22 cm x 28 cmPack of 100$11.15: 6264405
- Graph paper, polar coordinates, 22 cm x 28 cm-: -