Order Entry
United States
ContactUsLinkComponent
Glass Buret Funnel
Glass Buret Funnel
Catalog # 470235-710
Supplier:  GSC International
undefined
Glass Buret Funnel
Catalog # 470235-710
Supplier:  GSC International
Supplier Number:  205-5-CS

Specifications

  • Description:
    Buret Funnel
  • Environmentally Preferable:
  • Stem Length:
    36 mm
  • Stem Ext.∅:
    10 mm
  • Cat. No.:
    470235-710
  • Top diameter:
    40 mm

Specifications

About this item

Standard for any laboratory.

  • Made of borosilicate glass