Frog Development Slide Set
Frog Development Slide Set, 15, Cellular Development
Catalog # 470183-568
Supplier: Avantor
Jump to:
Product Details & Documents
Examples of Various Stages of Frog Development
- Identifying features clearly distinguishable
- Rigorous quality control standards
Ordering information: Set includes Frog Ovary (sect); Frog Testis (cs); Frog Sperm (sm); Unsegmented Egg (sect); Early Cleavage (sect); Late Cleavage (sect); Blastula (sect); Yolk Plug (sect); Gastrula (sect); Neural Plate (sect); Neural Fold (sect); Neural Tube (sect); 4mm Embryo (wm); 7mm Embryo (wm); and 10mm Embryo (wm).
Specifications
- Type:Cellular Development
- Description:Frog Development Slide Set
- No. of slides:15