Freeze-Dried Sheep Heart with Aorta and Trachea
Catalog # 470027-870
Supplier: Avantor
Specifications
- Description:Freeze-Dried Sheep Heart with Aorta and Trachea, Preserved, Each
- Cat. No.:470027-870
- Supplier no.:WARD470027-870
Specifications
About this item
Freeze-Dried Preserved.
This detailed preparation, for demonstrations or up-close study, includes the basic structures of the heart, as well as the aorta and trachea. The freeze-drying process uses no chemicals or fluids, making the specimen easier to handle; it can be rehydrated by following the included instructions.
Note: Ward's® preserved sheep materials is a byproduct of the food industry. Sheep are not raised specifically for dissection. These animals are recycled" for educational use."