Order Entry
United States
Orders LinkContactUsLinkComponent
Formic Acid

Formic Acid

Supplier:   Ward's Science
Formic Acid
Supplier:  Ward's Science
Certificates
Formic Acid

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS Number: 64-18-6
Formula Weight: 46.03
Formula: HCOOH
Hazard Info: Corrosive
Density (g/mL): 1.213
Boiling Point (°C): 101
Freezing Point (°C): -5
Solubility: Water, Alcohol and Ether
Synonyms: Methanoic Acid, Formylic Acid
Shelf Life (months): 12
Storage: White

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Product Family Options

Product Information

  • Grade
    Size
    Chemical/physical specifications
    Availability
    Price
  • 470301-118
    Formic Acid, Solution
    Formic Acid, Solution
    Laboratory
    100 mL
    88%
    Log in for availability
    Log in for availability
    Each (100ml)
    Each (100ml) - $18.00
    $18.00
    Each (100ml)
       $18.00

    Supplier Number: FF0270-100ML
  • 470301-120
    Formic Acid, Solution
    Formic Acid, Solution
    Laboratory
    500 mL
    88%
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $37.50
    $37.50
    Each (500ml)
       $37.50

    Supplier Number: FF0270-500ML

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.