Specifications
- Description:Flame Salts Set
- Cat. No.:470110-174
- Supplier no.:SP-4700
Specifications
About this item
Deluxe Set Includes Six Chemicals
See and study the spectra of six different chemicals — barium, boron, calcium, lithium, potassium, and strontium — when you do flame tests with this set of salts. Set of glass screw-topped bottles includes test tube and nichrome wire for holding the salts in a flame.