Order Entry
United States
Orders LinkContactUsLinkComponent
Fixed Speed Centrifuge

Fixed Speed Centrifuge

Fixed Speed Centrifuge, 254W x 203H x 302L mm, 3400 rpm, 6 tubes (2 - 10 ml)
Catalog # 470003-274
Supplier #:  C806
Fixed Speed Centrifuge
Catalog # 470003-274
Fixed Speed Centrifuge
Fixed Speed Centrifuge

Fixed Speed Centrifuge, 254W x 203H x 302L mm, 3400 rpm, 6 tubes (2 - 10 ml)

Price
$612.80
Log in for availability
Each
Product Details & Documents

A Valuable Addition to Any Laboratory.

  • 30 minute timer with bell tone signal
  • Latching lid with Safety Switch
  • Easily accessible stainless steel rotor

Economical, reliable, and very quiet. Suction cup rubber feet ensure stability and reduce vibration. Centrifuge features all UL approved components.

Specifications
  • Description:
    Fixed Speed Centrifuge
  • Dimensions:
    254W x 203H x 302L mm
  • Max. speed:
    3400 rpm
  • Tube capacity:
    6 tubes (2 - 10 ml)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.