Fish and Onion Mitosis Slide
Catalog # 470182-556
Supplier: Avantor
Specifications
- Description:Fish & Onion Mitosis
- Environmentally Preferable:
- Cat. No.:470182-556
- Stain:Weigert’s and Eosin
- Supplier no.:WARD470182-556
- Orientation:Section
Specifications
About this item
Compare and contrast mitosis
- Identifying features clearly distinguishable
- Rigorous quality control standards
Composite of three sections of fish blastodisc and three sections of onion root tip. Demonstrates animal and plant mitosis.