Firefly Digital Video Dermatoscope
Supplier: Avantor
Certificates
About this item
Magnify and Wirelessly Transmit Images
Now students can easily view skin conditions on PC monitors and TVs with accuracy and precision. This dermatoscope magnifies and wirelessly transmits live digital video and images directly to a PC. With a resolution of 2.0 megapixels, native optical magnification of 15-50X, and digital magnification of 15-150X, students can see crisp, clear images in full detail. Dual lens assembly features three-layer glass and a 650nm cutoff. Wireless link transmits at 2.4GHz across 4 channels. Includes built-in 800mAh battery and USB connection.
Frequently Bought Together