Order Entry
United States
ContactUsLinkComponent
Fieldmaster Turbidity Tube
Fieldmaster Turbidity Tube
Catalog # 470021-244
Supplier:  Science First
undefined
Fieldmaster Turbidity Tube
Catalog # 470021-244
Supplier:  Science First
Supplier Number:  623-2050

Specifications

  • Ø:
    4.5 cm
  • Description:
    Fieldmaster turbidity tube
  • Length:
    1200 mm
  • Cat. No.:
    470021-244
  • Supplier no.:
    78-070

Specifications

About this item

Thsese tubes meets GLOBE requirements.

  • Clear polycarbonate construction
  • Secchi disk design on the bottom
  • Aligned with GLOBE protocols

Measure turbidity in streams where a Secchi disk cannot be used. Simply drain the tube with a hose and finger clamp.