Fever Scan Tape
Catalog # 470012-694
Supplier: Liquid Crystal Resources
Specifications
- Description:Thermometer, LCD Temperature Tape (Fever Scan)
- Cat. No.:470012-694
- Supplier no.:FT-55U
Specifications
About this item
Pressed against the forehead for 35 to 40 seconds, this fever tester indicates body temperature.
- Range: 95 – 104 °F