BoneClones® Fetal Skull
Catalog # 470030-002
Supplier: BONE CLONES HOLDINGS
Small Business Enterprise
Specifications
- Description:Fetal Skull
- Height:8.6 cm
- Length:11.4 cm
- Cat. No.:470030-002
- Width:8.7 cm
- Supplier no.:BC-182
- No. of parts:1
Specifications
About this item
Visual Supplement For Growth And Development Studies
Show students the characteristics of a skull during stages of prenatal development. This full term Bone Clone® fetal skull is modeled after a fetus at 40 1/2 weeks old.