ContactUsLinkComponent
Fasciola Life History Slide Set
Fasciola Life History Slide Set
  470183-544
 :  Avantor
undefined
Fasciola Life History Slide Set
  470183-544
 :  Avantor
  • Type:
    Animal
  • Description:
    Fasciola Life History Slide Set
  • No. of slides:
    6
  • Cat. No.:
    470183-544
  • Supplier no.:
    WARD470183-544
Complete Life Cycle of This Common Flatworm
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Explore the Life Cycle Fasciola spp.
 : Set includes Adult (wm), Eggs (wm), Miracidia (wm), Redia (wm), Cercaria (wm), and Metacercaria (wm).