ContactUsLinkComponent
Fan Cart
  470105-306
undefined
Fan Cart
  470105-306
 :  10-105
  • Description:
    Fan cart
  • Cat. No.:
    470105-306
  • Supplier no.:
    10-105
Produce constant acceleration with this fan cart.
  • Fan provides constant force, resulting in constant acceleration
  • Removable plate vividly demonstrates Newton's third law
Graphically illustrate uniform acceleration independent of gravity. A motor-driven propeller provides thrust to accelerate the cart along a smooth level surface.
Made of durable, lightweight plastic, the cart features a 1.5×12.5 cm propeller, four low friction wheels, and a 14×15 cm removable sail.
Powered by four AA batteries, purchased separately. Overall size: 11×19.5×11 cm.