ContactUsLinkComponent
EZ-Open Pail
EZ-Open Pail
  470002-088
 :  Avantor
undefined
EZ-Open Pail
  470002-088
 :  Avantor
  • Capacity:
    13.2 L (3.5 gal.)
  • Color:
    White
  • Description:
    EZ-open pail
  • Cat. No.:
    470002-088
  • Supplier no.:
    WARD470002-088
EZ-Open pail with the gasketed lid.
  • Time saver and economical
  • Made of chemical-resistant polyethylene
  • Each pail has a plastic handle and a plastic hand grip
  • Aligned with GLOBE protocols
With just a few twists of wrist, lid can easily unscrewed and open these pails without needing a pail opener. Once after placing specimens in the pail, the lid can be tightly resealed to protect the contents for long-term storage.
Pail is labeled with the Ward’s Science logo.