Extra Long Nichrome Inoculating Needle
Catalog # 470101-678
Supplier: TONAWANDA MANUFACTURING
Specifications
- Description:Needle, Inoculating, Extra Long Nichrome Wire, 10 cm, Aluminum Handle
- Cat. No.:470101-678
- Supplier no.:4692900
Specifications
About this item
The 10cm wire makes it easy to reach cultures even in large culture tubes. With safety always in mind, its length also means easy, and safe, flame sterilizing.