Orders LinkContactUsLinkComponent
Essential Physics Demo: Spring Scale
Essential Physics Demo: Spring Scale
Essential Physics Demo: Spring Scale
Catalog # 470125-574
Supplier:   Wards
Supplier #:  1815-98
Essential Physics Demo: Spring Scale
Catalog # 470125-574
Supplier:  Wards
Essential Physics Demo: Spring Scale
Essential Physics Demo: Spring Scale
Essential Physics Demo: Spring Scale
Clearance
$71.65
Sale
Valid until 31/12/2026
Log in for availability
Each
Product Details & Documents
Affordable, Easy, Fun
Present these demonstrations in front of your class to excite your students about learning core physics concepts. Set up and execution are simple, so you can spend more time engaging your students. Ideal for introducing a unit or using as reinforcement after a lesson, these demos are available for all fundamental physical science topics.
Two 70 cm long spring scales allow students to easily see and demonstrate force experiments and equilibrium.
When a mass is hung midway between the two springs it causes equal displacement in each spring. If the mass is hung off–center, the two springs are elongated different distances. Displacement measurements can be easily assessed using the metric scale on the tube. Includes two large spring scales, a connecting rope with hook, and a fully–detailed demonstration guide. Each spring has a constant of approximately 78 N/m. Masses are not included.
Specifications
  • Description:
    Essential Physics Demo: Spring Scale