ContactUsLinkComponent
Essential Physics Demo: Newton Paradox
Essential Physics Demo: Newton Paradox
  470128-402
 :  Avantor
Small Business Enterprise
undefined
Essential Physics Demo: Newton Paradox
  470128-402
 :  Avantor
Small Business Enterprise
  • Description:
    Essential Physics Demo: Newton Paradox
  • Cat. No.:
    470128-402
  • Supplier no.:
    180874
A Puzzling Introduction to Inertia
Affordable, Easy, Fun
Present these demonstrations in front of your class to excite your students about learning core physics concepts. Set up and execution are simple, so you can spend more time engaging your students. Ideal for introducing a unit or using as reinforcement after a lesson, these demos are available for all fundamental physical science topics.
Challenge your students’ critical thinking skills with this intriguing demonstration that investigates the concepts of free–fall, inertia, and velocity. Drop two rods of unequal mass from the same height and show how they reach the ground at the same time. Then, ask students to predict the next outcome before launching the same rods downward to the ground simultaneously using a projectile launcher. Includes a launcher with rods of different masses and an instruction guide.