Equisetum
Equisetum
Catalog # 470176-288
Supplier: WATERSCAPES AQUATIC NURSERY
Supplier #: 865300
Jump to:
Product Details & Documents
Scouring Rush. It features reedy, upright growth habit and whorls of leaves at the nodes. The shoots, which contain silica, were once used to scrub utensils and pots, giving Equisetum its common name of scouring rush. It is supplied in a plastic bag with care instructions. Quantity: 8–12 shoots.
Documents
Mosses__Liverworts_Ferns
Product Literature
Specifications
- Description:Equisetum