Equilateral Refraction Prism
470021-796
- Description:Equilateral Refraction Prism
- Cat. No.:470021-796
- Supplier no.:FGP075
Demonstrate the bending properties of light.
- Clear flint glass
- Demonstrate refraction and reflection
This flint glass prism measures 3" long by 3/8" thick.