Endolimax nana, Trophozoites and Cysts Slide
470181-864
: Avantor
- Description:Endolimax nana, Trophozoites & Cysts
- Cat. No.:470181-864
- Stain:Hematoxylin
- Supplier no.:WARD470181-864
- Orientation:Smear
Intestinal Parasite
- Identifying features clearly distinguishable
- Rigorous quality control standards
Shows nuclei, small irregular deposits of chromatin, and large, acentric, irregularly-shaped karyosome.