ContactUsLinkComponent
Electrolysis Package
Electrolysis Package
Catalog # 470112-480
undefined
Electrolysis Package
Catalog # 470112-480
Supplier Number:  3556
Specifications
  • Description:
    Electrolysis Package
  • Cat. No.:
    470112-480
  • Supplier no.:
    3556
Specifications
About this item
This basic set includes: two heavily insulated, stainless steel electrodes; two clip-on wire connecting leads; and two rubber gaskets that provide a positive seal to inverted test tubes in a beaker.
  • Comes with instructions
Ordering information: Glassware is not supplied.