Eggs of Parasitic Worms Slide Set
470183-540
: Avantor
- Description:Eggs of Parasitic Worms Slide Set
- Environmentally Preferable:
- No. of slides:12
- Cat. No.:470183-540
- Supplier no.:WARD470183-540
Assortment of Eggs Belonging to the Most Prolific Parasites of the Mammalian Digestive Tract
- Identifying features clearly distinguishable
- Rigorous quality control standards
All specimens are prepared as whole mounts.
: Set includes Ascaris lumbricoides; Clonorchis sinensis; Diphyllobothrium latum; Enterobius vermicularis; Fasciola hepatica; Hookworm; Toxocara canis; Schistosoma haematobium; Haemonchus contortus; Schistosoma mansoni;
Taenia; and Trichuris trichiura.
Taenia; and Trichuris trichiura.