ContactUsLinkComponent
Ward's® Ecosystems in Distress: Oil Spill Cleanup Lab Activity
Ward's® Ecosystems in Distress: Oil Spill Cleanup Lab Activity
Catalog # 470030-194
Supplier:  Avantor
undefined
Ward's® Ecosystems in Distress: Oil Spill Cleanup Lab Activity
Catalog # 470030-194
Supplier:  Avantor
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Ward's® Ecosystems in Distress: Oil Spill Cleanup Lab Activity
  • No. of lab groups:
    6
  • Cat. No.:
    470030-194
Specifications
About this item
A Comprehensive, Hands-On Evaluation of Mechanical and Bioremedial Methods
  • Demonstrate the environmental impact of oil spills
  • Linked to a real world scenario
  • Includes a teacher's guide
Engage students in addressing real-world issues through an investigation covering environmental science, biotechnology, and socio-economics.
In this lab students will simulate miniature oil spills with the included materials and attempt various methods of cleanup, including skimming, absorbents, and oil-degrading microorganisms.
They can then compare and contrast the effectiveness of each approach, weighing it against financial and environmental costs. The activity contains a teacher’s guide, student copymaster, and materials for six setups.
Note: Includes perishable materials. Please specify delivery date