Economy Light Source for Optical Bench
Catalog # 470102-470
Supplier: United Scientific Supplies
Specifications
- Description:Light Source, Economy, for Optical Bench
- Cat. No.:470102-470
- Supplier no.:OBLS01
Specifications
About this item
Add a regular bulb as your light source for optical benches.
- Fits all standard meter sticks
- Provides consistent light
This economy light source is an excellent alternative to more expensive light sources. Wiring and power source are not included. Needs to be wired to a battery to work. Uses a miniature lamp.