ContactUsLinkComponent
Economy Alcohol Lamp Stand
Economy Alcohol Lamp Stand
Catalog # 470019-486
undefined
Economy Alcohol Lamp Stand
Catalog # 470019-486
Supplier Number:  ABSP05
Specifications
  • Description:
    Economy alcohol lamp stand
  • Cat. No.:
    470019-486
  • Supplier no.:
    ABSP05
Specifications
About this item
This rugged wire support stand has a heavy wire gauze top.
  • Wire gauze surface measures 8×10 cm (3×2")
  • Height: 11.5 cm (4½")
Made especially for alcohol burners.