ContactUsLinkComponent
Vernier EasyLink® Interface
Vernier EasyLink® Interface
Catalog # 470017-138
undefined
Vernier EasyLink® Interface
Catalog # 470017-138
Supplier Number:  EZ-LINK
Specifications
  • Description:
    Vernier® EasyLink® interface
  • Cat. No.:
    470017-138
  • Supplier no.:
    EZ-LINK
Specifications
About this item
Inexpensive data collection direct to any TI-Nspire® handheld or TI-84 Plus graphing calculator.
  • Connects BTA (sensor) to mini-A USB (calculator)
  • Compatible with all models of TI-84 Plus, TI-84 Plus CE, and TI-Nspire® CXII Texas Instruments calculators
  • Supports over 62 analog Vernier sensors (Digital sensors and microphone are incompatible)
EasyLink® is a single-channel interface that plugs into the USB port of a TI-84 Plus graphing Calculator or TI-Nspire® Handheld. Use EasyLink® and a supported sensor to explore math and science concepts found in real-world phenomena such as light, pressure, force, and more.