ContactUsLinkComponent
Earth Science Student Collection
  470104-714
 :  Avantor
undefined
Earth Science Student Collection
  470104-714
 :  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Earth Science Student Collection
  • Environmentally Preferable:
  • Cat. No.:
    470104-714
  • Supplier no.:
    WARD470104-714
Designed for high school and elementary college courses, these rock and mineral specimens provide a complete range of materials for comparisons of igneous, sedimentary and metamorphic rocks.
  • All specimens are numbered
  • Size of each: 2.5×5 cm
  • Set of 72
In addition, students can study rock-forming ores of metals and non-metals, differences in the hardness scale and physical properties including streak, luster, cleavage, fracture, specific gravity and magnetism. The collection is housed in two 36-compartment collection boxes, one each for rocks and minerals.
 : Includes streak plate, magnet, magnifier, identification list and instruction booklet.
 : Note: Collections are subject to change depending upon availability of individual items. Due to the nature of this product it may contain asbestos.