ContactUsLinkComponent
DNA Fingerprinting: A Simple Simulation
DNA Fingerprinting: A Simple Simulation
Catalog # 470004-150
Supplier:  Avantor
undefined
DNA Fingerprinting: A Simple Simulation
Catalog # 470004-150
Supplier:  Avantor
Specifications
  • Description:
    DNA Finger Printing: A Simple Simulation
  • Cat. No.:
    470004-150
  • Supplier no.:
    WARD470004-150
Specifications
About this item
A clear and easy way to show students the steps of DNA fingerprinting by simulation, this kit with accompanying DVD guides students through the step by step process: from gel to Southern blot to radioactive probe to fingerprint using everyday UPC codes.
  • Grades 10 to 12
  • Developed by Julie Bohrer
Students can then compare suspects with crime scene DNA.
Delivery: Includes materials for 15 student groups with DVD and instructions.