Depth-of-Field Practice Slide Set
470177-710
: Avantor
- Description:Depth-of-field practice slide set
- No. of slides:6
- Cat. No.:470177-710
- Supplier no.:WARD470177-710
Help Students Enhance Microscope Focusing Skills.
- Identifying features clearly distinguishable
- Rigorous quality control standards
: Each whole mount slide contains a different combination of intersecting, differently colored teased silk fibers. Students must correctly focus the high-power lens to determine the order in which they are mounted.