Den-Test Kit
Den-Test Kit
Catalog # 470007-790
Supplier: Kemtec
Supplier #: 1-110
Jump to:
Product Details & Documents
You don’t have to take a field trip to the dentist to teach your students about tooth decay and preventive oral hygiene. With this kit, students will gain firsthand knowledge as they culture bacteria taken from their own teeth both before and after brushing.
Includes materials for 24 students
sterile petri dishes
swabs
agar
mouthwash
complete instructions.
Documents
TDS Den-Test Kit
Datasheet
Specifications
- Description:Den-Test Kit