ContactUsLinkComponent
Density: Layers of Liquids Kit
Density: Layers of Liquids Kit
  470113-068
 :  Lab-Aids
undefined
Density: Layers of Liquids Kit
  470113-068
 :  Lab-Aids
 :  114
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Density: Layers of Liquids Student Activity
  • No. of lab groups:
    6
  • Cat. No.:
    470113-068
  • Includes:
    Teacher's Guide, Student Guides and Worksheets, SDS
  • Supplier no.:
    114
What Is Density?!
  • A substance has characteristic properties, which are independent of the amount of sample
  • A mixture of substances often can be separated into the original substances using one or more characteristic properties
  • Requires one 50 minute class period
Help students develop an operational definition of density. In this activity kit, liquids of different viscosity and density are provided, and students are asked to observe and predict, based on their observations, the relative density of each. They then test their predictions and develop an understanding of density by preparing sequences of liquid layers. When students are given values for the density of each liquid, the next challenge is to use this information to estimate the density of an unknown solid. Finally, there is an optional activity to design and carry out an experiment to determine the density of a fourth liquid.
 : Materials include Red Liquid, Green Liquid, Yellow Liquid, Density Tubes (LAB-AIDS®), End Caps for Density Tubes, Vial.