Cyclotella
Cyclotella Culture for 35 Students
Catalog # 470179-756
Supplier: Ward's Science
Supplier #: CALCYCLO
Jump to:
Product Details & Documents
Marine. Unicellular; small, centric cells. Marine medium.Characterized by shells (tests) made of silica, in two parts.
Documents
Algae_and_Cyanobacteria Working_with_Algae_and_Cyanobacteria
Product Literature
Specifications
- Description:Cyclotella Culture for 35 Students