ContactUsLinkComponent
Conductivity of Solutions Experiment
Catalog # 470015-250
Supplier:  Lab-Aids
undefined
Conductivity of Solutions Experiment
Catalog # 470015-250
Supplier:  Lab-Aids
Supplier Number:  313
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Conductivity of Solutions Experiment
  • No. of lab groups:
    15
  • Cat. No.:
    470015-250
  • Includes:
    Teacher Guide, SDS, Student Guide, Worksheets
  • Supplier no.:
    313
Specifications
About this item
This kit includes a series of fun activities that lead to a better understanding of the concept of ionization.
  • Study principles of ionization
  • Understand strong and weak electrolytes
  • Differentiate between strong and weak acids and bases
  • Requires one 50 minute class period to complete
Using LAB-AIDS® Conductivity Indicators, students study the principles of ionization by differentiating between strong electrolytes, weak electrolytes and non-electrolytes. They also learn to differentiate between strong and weak acids and strong and weak bases. Performing a series of fun activities, students observe and discover dramatic results which lead to a better understanding of the principles of ionization. This Lab-Aid is designed for use by students in introductory chemistry courses and higher.
Ordering information: Kit includes Chemplates® with measuring spatula, Ethyl alcohol, Hydrochloric acid, Hydrochloric acid, Ammonia, Glacial acetic acid, Deionized water, Anhydrous ethyl alcohol, Sodium chloride crystals, Sodium hydroxide pellets, and Sucrose crystals. Conductivity indicators are required for these activities but are not included.