ContactUsLinkComponent
Compressed Air Dispenser
Compressed Air Dispenser
Catalog # 470146-262
Supplier:  STONER
undefined
Compressed Air Dispenser
Catalog # 470146-262
Supplier:  STONER
Supplier Number:  94203CSTON01
Specifications
  • Description:
    Compressed air dispenser, 12 oz.
  • Environmentally Preferable:
  • Cat. No.:
    470146-262
  • Supplier no.:
    94203
Specifications
About this item
Ozone safe compressed air dispenser
Concentrated, directed jets of moisture-free filtered air safely blow dust particles from delicate instruments.