ContactUsLinkComponent
Complete Physical Geology Teaching Series
Complete Physical Geology Teaching Series
Catalog # 470025-366
Supplier:  Avantor
undefined
Complete Physical Geology Teaching Series
Catalog # 470025-366
Supplier:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Complete Physical Geology Teaching Series
  • Cat. No.:
    470025-366
Specifications
About this item
Buy the Complete Series and Save!
This complete teaching series contains 12 of the 15 topic and reference sets that cover every topic in the Physical Geology Series, including all tests students need to perform to identify rocks and minerals. This series contains the Physical Geology Reference Set, Hardness Topic Set, Cleavage, Fracture and Parting Topic Set, Streak, Color and Luster Topic Set, Tenacity and Special Properties Topic Set, Crystal Form Topic Set, Rock-Forming Minerals Topic Set, Igneous Rocks Topic Set, Sedimentary Rocks Topic Set, Metamorphic Rocks Topic Set, Metallic Resources Topic Set and the Nonmetallic Resources Topic Set.
The Classroom Bundles, which provide 15 bulk-packaged samples included in the sets and 15 sets of any required testing materials included with the corresponding set
*Classroom Bundles are bulk-packaged and do not include display boxes, or teacher/student activity guides.
Ordering information: Igneous, Sedimentary, and Metamorphic GEO-logic Thin Section Sets are not included.
Caution: Due to the nature of this product it may contain asbestos.