ContactUsLinkComponent
Ward's® College Algae Set
  470179-952
 :  Avantor
undefined
Ward's® College Algae Set
  470179-952
 :  Avantor
  • Description:
    WARD’S College Algae Set
  • Cat. No.:
    470179-952
  • Supplier no.:
    WARD470179-952
Compiles Types Most Often Studied at the University Level
Our comprehensive set includes algae usually featured in college textbooks. The 25 cultures include Anabaena, Bactrachospermum, Bangia, Chlamydomonas, Chlorella, Dictyota, Ectocarpus, Euglena, Fischerella, Gloeocapsa, Gleotrichia, Micrasterias, Netrium, Oedogonium, Oscillatoria, Peridinium, Scenedesmus, Spirogyra, Spirulina, Nitzschia, Synura, Ulothrix, Ulva, Vaucheria, and Volvox. The set also comes with the Working with Algae manual and materials for 35 students.