ContactUsLinkComponent
Coliscan® EasyGel® Water Monitoring Kit
Coliscan® EasyGel® Water Monitoring Kit
  470107-534
undefined
Coliscan® EasyGel® Water Monitoring Kit
  470107-534
 :  CWM10
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Coliscan® EasyGel® Water Monitoring Kit
  • No. of lab groups:
    10
  • Cat. No.:
    470107-534
  • Supplier no.:
    CWM10
Make Sure the Water is Free of E. coli
  • Collect local water samples
  • Linked to a real world scenario
  • Includes an instruction guide
Coliscan Water Monitoring Kit features Easygel, an agar replacement. Each kit has materials for 10 tests. The kit includes Coliscan Easygel bottles, Easygel petri dishes, 30mL collection bottles, 1mL Droppers, instruction guide, and Coliscan Interpretation Guide.