Ward's® Coliphage Culture and Titer Determination Lab Activity
Catalog # 470030-364
Supplier: Avantor
Specifications
- Description:Coliphage Culture and Titer Determination Kit
- No. of lab groups:6
- Cat. No.:470030-364
Specifications
About this item
Produce High-Titer T-Series Coliphages Without Setting Up an Aeration System
- Practice aseptic technique
- Demonstrate cell damage caused by viral infection
- Materials for six lab groups
- Time Required: ~90 minutes over 3 days
Introduce students to the lytic mode of a bacteriophage life cycle. Or you can use the serial dilution technique to explore the concept of bacteriophage titer, which is the concentration of infectious virus particles per milliliter of suspension fluid. The simple procedure involves a serial dilution of the bacteriophage stock followed by the double agar overlay technique. The kit comes with all necessary materials including E. coli B and coliphage T4r cultures, and study guide.
Ordering information: Coupon is included for redemption of perishable materials. Redeem by mail, phone, fax, or email.