ContactUsLinkComponent
Coliform in Water Test Lab Activity
Coliform in Water Test Lab Activity
Catalog # 470193-380
Supplier:  MICROLOGY INC
undefined
Coliform in Water Test Lab Activity
Catalog # 470193-380
Supplier:  MICROLOGY INC
Supplier Number:  K24WQ
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Coliform in Water Test Lab Activity
  • No. of lab groups:
    24
  • Cat. No.:
    470193-380
  • Supplier no.:
    K24WQ
Specifications
About this item
Students apply lab work to real-life issues.
  • Students Collect Local Water Samples and Tests for Coliform Bacteria
  • Test Results Compared to Safe and Unsafe Quantities
  • Sewage Treatment Facilities Test for the Same Bacterial Group
  • Links to Real-World Scenarios
  • Teacher's Manual Included
They identify a bacterial group for which actual sewage treatment facilities test.
The class samples local water for coliform bacteria then compares results to a chart showing safe and unsafe quantities.
Ordering information: This kit includes instruction and material for a class of 24 students including Easygel, a pre-measured, sterilized culture media that requires no mixing or autoclaving.
Delivery: This kit includes perishable materials. Please specify delivery date.