Coleus, Stem Tip Slide
Catalog # 470181-316
Supplier: Avantor
Specifications
- Description:Coleus, Stem Tip Slide
- Environmentally Preferable:
- Cat. No.:470181-316
- Stain:Quadruple
- Supplier no.:WARD470181-316
- Orientation:Longitudinal section
Specifications
About this item
The Coleus Stem Tip Slide demonstrates leaf development.
- Identifying features clearly distinguishable
- Rigorous quality control standards
Shows apical meristem and developing leaves.