Coccolithophora
Coccolithophora Culture for 35 Students
Catalog # 470179-912
Supplier: Ward's Science
Supplier #: COCCO30/35
Jump to:
Product Details & Documents
Marine. Unicellular; contains same pigments as other Golden Algae but contains haptonema, unique to Haptophyceae. Coccoid, from coccoliths. Marine medium. Characterized by golden yellow plastids.
Documents
Algae_and_Cyanobacteria Working_with_Algae_and_Cyanobacteria
Product Literature
Specifications
- Description:Coccolithophora Culture for 35 Students