ContactUsLinkComponent
Classification of Matter Kit
Classification of Matter Kit
Catalog # 470004-146
Supplier:  Avantor
undefined
Classification of Matter Kit
Catalog # 470004-146
Supplier:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Classification of Matter Kit
  • No. of lab groups:
    30
  • Cat. No.:
    470004-146
  • Supplier no.:
    WARD470004-146
Specifications
About this item
Learn how to use a dichotomous key.
  • Ideal for grades 5-8
  • Learn to classify materials based on their innate characteristics
  • Includes materials for 30 students
This dichotomous key set helps students to learn about the logical method of using a dichotomous key to classify matter by properties. This activity provides students with questions that will help them to classify samples of matter into specific groups. Designed by Texas chemistry teachers Alicia Silva and Frances Sprabary. Topics covered include energy mixtures, pure substances, colloids, suspensions, solutions, elements and compounds.
Ordering information: The kit includes aquarium gravel, sugar, salt water, sulfur, lead shot, glass and delrin beads, milk of magnesia, sand, copper turnings, water, blue-colored water, glue. Includes teacher's guide and student copymaster. Required but not provided: Large test tubes (optional), cider vinegar (optional), food coloring (optional), flashlight (optional).